Would you like to log in again?
Tubing adapters
Supplier: Smiths Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
229-0021EA 9.9 GBP InStock
229-0021 229-0020 229-0019 229-0018
Tubing adapters
Fittings and Connectors
Nylon, white
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Withstand sterilisation to +136 °C
The diameters shown are minimum to maximum for each end.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty